Learn More
Biomatik Corporation Human ON (Osteonectin) ELISA Kit
Supplier:  Biomatik Corporation EKF57673
Encompass_Preferred
Human ON (Osteonectin) ELISA Kit for detecting ON in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P09486. Target Synonyms: ON; BM-40; Basement-membrane protein 40; BM-40; ONcysteine-rich protein; Secreted protein acidic and rich in cysteine; secreted protein; acidic; cysteine-rich; osteonectin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5447