Learn More
Biomatik Corporation Human PGAM1 (PhosphoglyceRate Mutase 1) ELISA Kit
Supplier:  Biomatik Corporation EKF57684
Encompass_Preferred
Human PGAM1 (PhosphoglyceRate Mutase 1) ELISA Kit for detecting PGAM1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.781-50ng/ml. Sensitivity: 0.469ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P18669. Target Synonyms: PGAM1; CDABP0006; PGAMA; PGAM-B; BPG-dependent PGAM 1; PhosphoglyceRate mutase isozyme B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5452