Learn More
Biomatik Corporation Rat IL-5 (Interleukin-5) ELISA Kit
Supplier:  Biomatik Corporation EKF57867
Encompass_Preferred
Rat IL-5 (Interleukin-5) ELISA Kit for detecting IL-5 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q08125. Target Synonyms: IL-5; IL5; EDF; TRF; E-CSF; ESGF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5467