Learn More
Biomatik Corporation Rat MMP-1 (Matrix Metalloproteinase 1) ELISA Kit
Supplier:  Biomatik Corporation EKF57942
Encompass_Preferred
Rat MMP-1 (Matrix Metalloproteinase 1) ELISA Kit for detecting MMP-1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P81563. Target Synonyms: MMP-1; Matrix Metalloproteinase 1; CLGmatrix metalloprotease 1; CLGN; Fibroblast collagenase; interstitial collagenase; Matrix metalloproteinase-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5472