Learn More
Biomatik Corporation Rat ENG (Endoglin) ELISA Kit
Supplier:  Biomatik Corporation EKF57971
Encompass_Preferred
Rat ENG (Endoglin) ELISA Kit for detecting ENG in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 62.5-4000pg/ml. Sensitivity: 37.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q6Q3E8. Target Synonyms: Endoglin; CD105; ENG; END; HHT1; ORW; ORW1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5476