Learn More
Biomatik Corporation Rat GS (Glutamine synthetase) ELISA Kit
Supplier:  Biomatik Corporation EKF58010
Encompass_Preferred
Rat GS (Glutamine synthetase) ELISA Kit for detecting GS in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P09606. Target Synonyms: GS. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5485