Learn More
Biomatik Corporation Rat Comp (Cartilage oligomeric matrix protein) ELISA Kit
Supplier:  Biomatik Corporation EKF58036
Encompass_Preferred
Rat Comp (Cartilage oligomeric matrix protein) ELISA Kit for detecting Comp in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P35444. Target Synonyms: Comp; EDM1; EPD1; MED; PSACH; Thrombospondin-5; cartilage oligomeric matrix protein; pseudoachondroplasia; epiphyseal dysplasia1; multiple; cartilage oligomeric matrix protein; pseudoachondroplasia; epiphyseal dysplasia1; multiple; EDM1; EPD1; MEDMGC131819; MGC149768; PSACH; pseudoachondroplasia; epiphyseal dysplasia 1; multiple; THBS5; Thrombospondin-5; thrombospondin-5; TSP5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5499