Learn More
Biomatik Corporation Mouse CORT (Corticosterone) ELISA Kit
Supplier:  Biomatik Corporation EKF58040
Encompass_Preferred
Mouse CORT (Corticosterone) ELISA Kit for detecting CORT in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 2.813-180ng/ml. Sensitivity: 1.688ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: CORT. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5501