Learn More
Biomatik Corporation Mouse PAP (Plasmin-Antiplasmin Complex) ELISA Kit
Supplier:  Biomatik Corporation EKF58048
Encompass_Preferred
Mouse PAP (Plasmin-Antiplasmin Complex) ELISA Kit for detecting PAP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q61247. Target Synonyms: PAP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5505