Learn More
Biomatik Corporation Human S100A4 (Protein S100-A4) ELISA Kit
Supplier:  Biomatik Corporation EKF58068
Encompass_Preferred
Human S100A4 (Protein S100-A4) ELISA Kit for detecting S100A4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P26447. Target Synonyms: S100A4; Protein S100-A4; S100 calcium-binding protein A4; Protein Mts1; Placental calcium-binding protein; Calvasculin; Metastasin; CAPL; MTS1; 42A; CAPL; MTS1; P9KA; PEL98; 18A2; CAPLS100 calcium binding protein A4; fibroblast-specific protein-1; 42A; FSP1; leukemia multidrug resistance associated protein; malignant transformation suppression 1; murine placental homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5516