Learn More
Biomatik Corporation Human LRRK2 (Leucine-rich repeat serine/threonine-protein kinase 2) ELISA Kit
Supplier:  Biomatik Corporation EKF58070
Encompass_Preferred
Human LRRK2 (Leucine-rich repeat serine/threonine-protein kinase 2) ELISA Kit for detecting LRRK2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q5S007. Target Synonyms: LRRK2; AURA17; Dardarin; PARK8; ROCO2; augmented in rheumatoid arthritis 17; leucine-rich repeat kinase 2; leucine-rich repeat serine; threonine-protein kinase 2; RIPK7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5518