Learn More
Biomatik Corporation Mouse C-Peptide (C-Peptide) ELISA Kit
Supplier:  Biomatik Corporation EKF58072
Encompass_Preferred
Mouse C-Peptide (C-Peptide) ELISA Kit for detecting C-Peptide in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P01325; P01326. Target Synonyms: C-P; C-Peptide; Connecting Peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5520