Learn More
Biomatik Corporation Human GPA33 (Cell surface A33 antigen) ELISA Kit
Supplier:  Biomatik Corporation EKF58090
Encompass_Preferred
Human GPA33 (Cell surface A33 antigen) ELISA Kit for detecting GPA33 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q99795. Target Synonyms: GPA33; A33; Glycoprotein A33; glycoprotein A33; transmembrane; transmembrane glycoprotein A33. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5533