Learn More
Biomatik Corporation Rat Apln (Apelin) ELISA Kit
Supplier:  Biomatik Corporation EKF58098
Encompass_Preferred
Rat Apln (Apelin) ELISA Kit for detecting Apln in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: Q9R0R3. Target Synonyms: Apln; Apelin; APEL; XNPEP2; APJ endogenous ligand. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5539