Learn More
Biomatik Corporation Human AFP-L3 (Alpha-fetoprotein Lens culinaris agglutiin 3) ELISA Kit
Supplier:  Biomatik Corporation EKF58100
Encompass_Preferred
Human AFP-L3 (Alpha-fetoprotein Lens culinaris agglutiin 3) ELISA Kit for detecting AFP-L3 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.375ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P02771. Target Synonyms: AFP-L; Alpha-fetoprotein Lens culinaris agglutiin 3; AFPL3; aFPL3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5541