Learn More
Biomatik Corporation Mouse PPAR-γ (Peroxisome ProlifeRator-activated receptor γ) ELISA Kit
Supplier:  Biomatik Corporation EKF58106
Encompass_Preferred
Mouse PPAR-γ (Peroxisome ProlifeRator-activated receptor γ) ELISA Kit for detecting PPAR-γ in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P37238. Target Synonyms: PPAR-γ; Peroxisome ProlifeRator Activated Receptor Gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5547