Learn More
Biomatik Corporation Human WWC1 (Protein KIBRA) ELISA Kit
Supplier:  Biomatik Corporation EKF58112
Encompass_Preferred
Human WWC1 (Protein KIBRA) ELISA Kit for detecting WWC1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8IX03. Target Synonyms: WWC1; KIAA0869; KIBRA; WW domain-containing protein 1; HBeAg-binding protein 3; Kidney and brain protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5551