Learn More
Biomatik Corporation Human CXCR5 (C-X-C chemokine receptor type 5) ELISA Kit
Supplier:  Biomatik Corporation EKF58127
Encompass_Preferred
Human CXCR5 (C-X-C chemokine receptor type 5) ELISA Kit for detecting CXCR5 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P32302. Target Synonyms: CXCR5; C-X-C chemokine receptor type 5; CXC-R5; CXCR-5; Burkitt lymphoma receptor 1; Monocyte-derived receptor 15; MDR-15; BLR1; CD185. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5565