Learn More
Biomatik Corporation Human IL-31 (Interleukin-31) ELISA Kit
Supplier:  Biomatik Corporation EKF58149
Encompass_Preferred
Human IL-31 (Interleukin-31) ELISA Kit for detecting IL-31 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.813-500pg/ml. Sensitivity: 4.688pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q6EBC2. Target Synonyms: IL-31. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5578