Learn More
Biomatik Corporation Human TEX101 (testis expressed 101) ELISA Kit
Supplier:  Biomatik Corporation EKF58161
Encompass_Preferred
Human TEX101 (testis expressed 101) ELISA Kit for detecting TEX101 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9BY14. Target Synonyms: Cell surface receptor NYD SP8; GTPR867; NYD SP8; PRO1884; SGRG; TES101RP; testis expressed 101; Testis expressed protein 101; TEX101. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5588