Learn More
Biomatik Corporation Mouse MUP20 (Major urinary protein 20) ELISA Kit
Supplier:  Biomatik Corporation EKF58167
Encompass_Preferred
Mouse MUP20 (Major urinary protein 20) ELISA Kit for detecting MUP20 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Accession Number: Q5FW60. Target Synonyms: MUP20; Darcin; Major urinary protein 24; ; Mup24. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5591