Learn More
Biomatik Corporation Rat SOD2 (Superoxide Dismutase 2, Mitochondrial) ELISA Kit
Supplier:  Biomatik Corporation EKF58184
Encompass_Preferred
Rat SOD2 (Superoxide Dismutase 2, Mitochondrial) ELISA Kit for detecting SOD2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P07895. Target Synonyms: SOD2; Superoxide dismutase [Mn]; mitochondrial; IPO-B; Mn SOD; SOD; Mitochodrial; indophenoloxidase B; IPOB; manganese-containing superoxide dismutase; mangano-superoxide dismutase; Mn superoxide dismutase; MNSOD; MVCD6; superoxide dismutase [Mn]; mitochondrial; superoxide dismutase 2; mitochondrial. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5602