Learn More
Biomatik Corporation Human dopamine, DA ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC33498
Encompass_Preferred
The Human dopamine, DA ELISA Kit allows for quantitative determination of dopamine, DA in serum, plasma, tissue homogenates and cell lysates. Species Reactivity: human. Detection Range: 0.5 ng/mL-100 ng/mL. Sensitivity: 0.25 ng/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-7853