Learn More
Biomatik Corporation Mouse GDF1 (Growth Differentiation Factor 1) ELISA Kit
Supplier:  Biomatik Corporation EKF58211
Encompass_Preferred
Mouse GDF1 (Growth Differentiation Factor 1) ELISA Kit for detecting GDF1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P20863. Target Synonyms: DORV; DTGA3; embryonic growth; differentiation factor 1; GDF-1; growth differentiation factor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5623