Learn More
Biomatik Corporation Mouse COL4 (Collagen Type IV) ELISA Kit
Supplier:  Biomatik Corporation EKF58219
Encompass_Preferred
Mouse COL4 (Collagen Type IV) ELISA Kit for detecting COL4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P08122; P02463; Q9QZR9; Q9QZS0. Target Synonyms: COL4; Type-IV Collagen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5629