Learn More
Biomatik Corporation Mouse FPR2 (Formyl Peptide Receptor 2) ELISA Kit
Supplier:  Biomatik Corporation EKF58230
Encompass_Preferred
Mouse FPR2 (Formyl Peptide Receptor 2) ELISA Kit for detecting FPR2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O88536. Target Synonyms: FPR2; RFP; Lipoxin A4 receptor; LXA4 receptor; HM63; FMLP-related receptor I; FMLP-R-I; Formyl peptide receptor-like 1; FPRH1; FPRL1; LXA4R. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5635