Learn More
Biomatik Corporation Human CD13 (Aminopeptidase N) ELISA Kit
Supplier:  Biomatik Corporation EKF58245
Encompass_Preferred
Human CD13 (Aminopeptidase N) ELISA Kit for detecting CD13 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P15144. Target Synonyms: CD13; AAP; Aminopeptidase N; ; ANPEP; APN; GP150; LAP1; P150; PEPN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5646