Learn More
Biomatik Corporation Mouse DBP/GC (Vitamin D-binding protein) ELISA Kit
Supplier:  Biomatik Corporation EKF58250
Encompass_Preferred
Mouse DBP/GC (Vitamin D-binding protein) ELISA Kit for detecting DBP; GC in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.906-250ng/ml. Sensitivity: 2.344ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P21614. Target Synonyms: DBP; DBP; GC; GRD3; VDBG; VDBP Vitamin D-binding alpha-globulin; vitamin D-binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5650