Learn More
Biomatik Corporation Mouse Cp (Ceruloplasmin) ELISA Kit
Supplier:  Biomatik Corporation EKF58268
Encompass_Preferred
Mouse Cp (Ceruloplasmin) ELISA Kit for detecting Cp in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 4.688-300ng/ml. Sensitivity: 2.813ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q61147. Target Synonyms: Cp. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5663