Learn More
Biomatik Corporation Human HCN4 (Potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 4) ELISA Kit
Supplier:  Biomatik Corporation EKF58272
Encompass_Preferred
Human HCN4 (Potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 4) ELISA Kit for detecting HCN4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9Y3Q4. Target Synonyms: HCN4; Potassium; sodium hyperpolarization-activated cyclic nucleotide-gated channel 4; hyperpolarization activated cyclic nucleotide-gated cation channel 4; hyperpolarization activated cyclic nucleotide-gated potassium channel 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5666