Learn More
Biomatik Corporation Mouse Chia (Acidic mammalian chitinase) ELISA Kit
Supplier:  Biomatik Corporation EKF58284
Encompass_Preferred
Mouse Chia (Acidic mammalian chitinase) ELISA Kit for detecting Chia in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q91XA9. Target Synonyms: Chia; Lung-specific protein TSA1902; AMCase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5675