Learn More
Biomatik Corporation Rat Map2 (Microtubule-associated protein 2) ELISA Kit
Supplier:  Biomatik Corporation EKF58292
Encompass_Preferred
Rat Map2 (Microtubule-associated protein 2) ELISA Kit for detecting Map2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P15146. Target Synonyms: Map2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5680