Learn More
Biomatik Corporation Mouse 8-iso-PGF2α (8-isoprostane) ELISA Kit
Supplier:  Biomatik Corporation EKF58312
Encompass_Preferred
Mouse 8-iso-PGF2α (8-isoprostane) ELISA Kit for detecting 8-iso-PGF2α in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Competitive ELISA, Coated with Antigen. Assay Time: 3h, 30 min. Target Synonyms: 8-isoprostane. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5692