Learn More
Biomatik Corporation Mouse Ahr (Aryl hydrocarbon receptor) ELISA Kit
Supplier:  Biomatik Corporation EKF58328
Encompass_Preferred
Mouse Ahr (Aryl hydrocarbon receptor) ELISA Kit for detecting Ahr in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.391-25ng/ml. Sensitivity: 0.234ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P30561. Target Synonyms: Ahr; ; Aryl hydrocarbon receptor; BHLHE76; Ah receptor; AH-receptor; aromatic hydrocarbon receptor; Class E basic helix-loop-helix protein 76. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5704