Learn More
Biomatik Corporation Rat SRD5A2 (5 alpha Reductase 2) ELISA Kit
Supplier:  Biomatik Corporation EKF58335
Encompass_Preferred
Rat SRD5A2 (5 alpha Reductase 2) ELISA Kit for detecting SRD5A2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P31214. Target Synonyms: 5 alpha Reductase 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5708