Learn More
Biomatik Corporation Mouse INSL3 (Insulin-Like 3) ELISA Kit
Supplier:  Biomatik Corporation EKF58342
Encompass_Preferred
Mouse INSL3 (Insulin-Like 3) ELISA Kit for detecting INSL3 in cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: O09107. Target Synonyms: INSL3; Insulin-like 3; Ley I-L; RLFley-I-L; RLF; RLNL; insulin-like 3; insulin-like 3; Leydig cell; leydig insulin -like hormone; leydig insulin -like peptide; Relaxin-like factor; relaxin-like factor b. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5711