Learn More
Biomatik Corporation Rat Cs (CitRate synthase, mitochondrial) ELISA Kit
Supplier:  Biomatik Corporation EKF58373
Encompass_Preferred
Rat Cs (CitRate synthase, mitochondrial) ELISA Kit for detecting Cs in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.125-200ng/ml. Sensitivity: 1.875ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8VHF5. Target Synonyms: Cs. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5735