Learn More
Biomatik Corporation Human Acetylated p53 (Acetylated p53) ELISA Kit
Supplier:  Biomatik Corporation EKF58386
Encompass_Preferred
Human Acetylated p53 (Acetylated p53) ELISA Kit for detecting Acetylated p53 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 46.875-3000pg/ml. Sensitivity: 28.125pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P04637. Target Synonyms: Acetylated p53. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5743