Learn More
Biomatik Corporation Mouse Arg1 (Arginase-1) ELISA Kit
Supplier:  Biomatik Corporation EKF58420
Encompass_Preferred
Mouse Arg1 (Arginase-1) ELISA Kit for detecting Arg1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.563-100miu/ml. Sensitivity: 0.938miu/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q61176. Target Synonyms: Arg1; ; Arginase-1; Liver Arginase; Liver-type arginase; Type I arginase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5764