Learn More
Biomatik Corporation Human ADA (Adenosine deaminase) ELISA Kit
Supplier:  Biomatik Corporation EKF58429
Encompass_Preferred
Human ADA (Adenosine deaminase) ELISA Kit for detecting ADA in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P00813. Target Synonyms: ADA; Adenosine Deaminase; ADA1; adenine deaminase; Adenosine aminohydrolase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5770