Learn More
Biomatik Corporation Human GSTP1 (Glutathione S-transferase P) ELISA Kit
Supplier:  Biomatik Corporation EKF58446
Encompass_Preferred
Human GSTP1 (Glutathione S-transferase P) ELISA Kit for detecting GSTP1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.344-150ng/ml. Sensitivity: 1.406ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P09211. Target Synonyms: GSTP1; GSTpi; Glutathione S Transferases Pi; DFN7; FAEES3; deafness; X-linked 7; GST class-pi; GST3; DFN7; GSTP; GSTP1-1; PI; fatty acid ethyl ester synthase III; glutathione S-transferase P; glutathione S-transferase pi 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5782