Learn More
Biomatik Corporation Rat DKK1 (Dickkopf Related Protein 1) ELISA Kit
Supplier:  Biomatik Corporation EKF58448
Encompass_Preferred
Rat DKK1 (Dickkopf Related Protein 1) ELISA Kit for detecting DKK1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.313-20ng/ml. Sensitivity: 0.188ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: G5CAC0. Target Synonyms: DKK1; dickkopf; Xenopus laevis homolog 1; dickkopf homolog 1; Xenopus laevis; dickkopf related protein-1; Dickkopf-1; dickkopf-related protein 1; Dkk-1; DKK-1; hDkk-1; SKdickkopf-1 like. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5784