Learn More
Biomatik Corporation Human NXPE4 (NXPE family member 4) ELISA Kit
Supplier:  Biomatik Corporation EKF58461
Encompass_Preferred
Human NXPE4 (NXPE family member 4) ELISA Kit for detecting NXPE4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q6UWF7. Target Synonyms: C11orf33; FAM55D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5793