Learn More
Biomatik Corporation Mouse NOS3/eNOS (Nitric Oxide Synthase 3, Endothelial) ELISA Kit
Supplier:  Biomatik Corporation EKF58481
Encompass_Preferred
Mouse NOS3/eNOS (Nitric Oxide Synthase 3, Endothelial) ELISA Kit for detecting NOS3; eNOS in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P70313. Target Synonyms: NOS3; eNOS; Constitutive NOS; ECNOS; endothelial; Endothelial NOS; eNOS; cNOS; nitric oxide synthase 3; endothelial cell; NOS type III; NOSIII. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5810