Learn More
Biomatik Corporation Rat MIγ/CXCL9 (Monocyte Interferon Gamma Inducing Factor) ELISA Kit
Supplier:  Biomatik Corporation EKF58530
Encompass_Preferred
Rat MIγ/CXCL9 (Monocyte Interferon Gamma Inducing Factor) ELISA Kit for detecting MIγ; CXCL9 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8K4B1. Target Synonyms: CXCL9; Miγ; MIG; CMK; crg-10; C-X-C motif chemokine 9; Gamma-interferon-induced monokine; Humig; HuMIG; MIGSmall-inducible cytokine B9; monokine induced by gamma interferon. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5844