Learn More
Biomatik Corporation Human DEFB105 (Beta-defensin 105) ELISA Kit
Supplier:  Biomatik Corporation EKF58533
Encompass_Preferred
Human DEFB105 (Beta-defensin 105) ELISA Kit for detecting DEFB105 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78.125-5000pg/ml. Sensitivity: 46.875pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q8NG35. Target Synonyms: Beta-defensin 105; Beta-defensin 5; BD-5; DEFB-5; Defensin, beta 105; BD5; DEFB105; DEFB5; AND; DEFB105B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5847