Learn More
Biomatik Corporation Mouse Fabp4 (Fatty acid-binding protein, adipocyte) ELISA Kit
Supplier:  Biomatik Corporation EKF58535
Encompass_Preferred
Mouse Fabp4 (Fatty acid-binding protein, adipocyte) ELISA Kit for detecting Fabp4 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P04117. Target Synonyms: A FABP; AFABP; ALBP; aP2; FABP4; Fatty acid binding protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5849