Learn More
Biomatik Corporation Rat Lp-a (Lipoprotein a) ELISA Kit
Supplier:  Biomatik Corporation EKF58540
Encompass_Preferred
Rat Lp-a (Lipoprotein a) ELISA Kit for detecting Lp-a in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.563-100ng/ml. Sensitivity: 0.938ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P04639. Target Synonyms: Lp-a; LPA; Lp; a; AK38; APOA; LP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5853