Learn More
Biomatik Corporation Human TSLP (Thymic stromal lymphopoietin) ELISA Kit
Catalog No. 502295862
Encompass_Preferred
Human TSLP (Thymic stromal lymphopoietin) ELISA Kit for detecting TSLP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q969D9. Target Synonyms: TSLP; Thymic stromal lymphopoietin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5862 Supplier Biomatik Corporation Supplier No. EKF58551
Human TSLP (Thymic stromal lymphopoietin) ELISA Kit for detecting TSLP in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q969D9. Target Synonyms: TSLP; Thymic stromal lymphopoietin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.