Learn More
Biomatik Corporation Human STUB1 (E3 ubiquitin-protein ligase CHIP) ELISA Kit
Supplier:  Biomatik Corporation EKF58582
Encompass_Preferred
Human STUB1 (E3 ubiquitin-protein ligase CHIP) ELISA Kit for detecting STUB1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9UNE7. Target Synonyms: STUB1; HSPABP2; NY-CO-7; SDCCAG7; STUB1; UBOX1; Antigen NY-CO-7; Carboxy terminus of Hsp70-interacting protein; CHIPSTIP1 homology and U box-containing protein 1; CLL-associated antigen KW-8; E3 ubiquitin-protein ligase CHIP; EC 6.3.2.-; heat shock protein A binding protein 2; c-terminal; HSPABP2; NY-CO-7; SDCCAG7; serologically defined colon cancer antigen 7; UBOX1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5884